AB-PINACA N-pentanoic acid metabolite-d4 (CRM), Cayman Chemical
Supplier: Cayman Chemical Company
18145-100
76543-974EA 96 USD
76543-974
AB-PINACA N-pentanoic acid metabolite-d4 (CRM), Cayman Chemical
Standards Organic Standards
An internal standard for the quantification of AB-PINACA N-pentanoic acid metabolite by GC- or LC-MS
  • This product is qualified as a Certified Reference Material (CRM) that has been manufactured and tested to meet ISO/IEC 17025 and ISO 17034 international standards
  • This CRM has been characterized by metrologically valid procedures and may be used as a quantitative analytical reference standard