Wedelolactone ≥98%

Supplier: Cayman Chemical Company

Synonyms: 1,8,9-Trihydroxy-3-methoxy-6H-benzofuro[3,2-c]chromen-6-one, 7-Methoxy-5,11,12-trihydroxy-coumestan

11796-500 11796-1 11796-5
76293-104EA 63.3 USD
76293-104 76293-106 76293-108
Wedelolactone ≥98%
Wedelolactone

  • A natural NF-κB inhibitor


A natural coumestan that inhibits NF-κB signaling at the level of IKK action, resulting in suppression of NF-κB-mediated gene expression at concentrations of 1-100 µM; also inhibits hepatitis C virus NS5B polymerase in vitro (IC50 = 36 µM), STAT1 dephosphorylation (50 µM), and EZH2-EED interactions (Kd = 2.8 µM)

Formula: C₁₆H₁₀O₇
Storage Temperature: Freezer
MDL Number: MFCD07778564
CAS Number: 524-12-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6d1271b247cda7a35e9d61d6460b5a40698411bd&vid=6d1271b247cda7a35e9d61d6460b5a40698411bd&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e706530caed47bda8aa4699651024f3dc7602ac2&vid=e706530caed47bda8aa4699651024f3dc7602ac2&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0b6bce559859b7d933903fced69314b258f4ec3a&vid=0b6bce559859b7d933903fced69314b258f4ec3a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Formulation A crystalline solid
Purity ≥98%
Stability ≥ 2 years


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR