Blade Sets, G2 Series, American Hakko Products
Supplier: AMERICAN HAKKO PRODUCTS INC TE
G2-A1602
76277-814PK 760.75 USD
76277-814
Blade Sets, G2 Series, American Hakko Products
Soldering Supplies Desoldering Tools
G2 series composite blades used with FT-801 thermal wire stripper.
With or without wishbone support between the blades that is attached to the collar. This support mitigates rotational play in the blades to improve notch alignment at the end of the blades when tighter tolerances are required.