Semco® Sealant Gun, Model 250-A, Bergdahl Associates
Supplier: BERGDAHL ASSOCIATES INC. TE
250255
76250-842EA 2990.39 USD
76250-842
Semco® Sealant Gun, Model 250-A, Bergdahl Associates
Soldering Supplies Desoldering Tools
The Semco® model 250-A sealant gun is the standard of the aerospace and electronics industries. This is constructed of durable metal and engineered plastic parts to reduces applied material cost.
  • Provides accurate material control
  • Bayonet lock on retainer permits quick cartridge change
  • Touch control on trigger regulates flow
  • Positive control for smooth, even dispensing
  • Ergonomic design and quiet operation
It is designed for quiet pneumatic application of sealants, adhesives, potting compounds, and other materials. The nickel-plated retainers are affixed to the valve assembly via a quick connect, secure bayonet lock fixture permitting easy replacement of cartridges.
Ordering information: Air hoses to operate the gun must be ordered separately.
Delivery information: The Semco® model 250-A gun is available with retainers to fit standard 32 cc, 59 cc, 156 cc, 214 cc, and 310 cc disposable cartridges.