Solder Pot Cartridge, JBC
Supplier: JBC TOOLS USA INC. TE
C245-SP01
76250-116EA 263.12 USD
76250-116
Solder Pot Cartridge, JBC
Soldering Supplies Desoldering Tools
This model is ideal for tinning wires and components faster and safer.
  • Can be used also to desolder through hole multipin components
The long-lasting coating pot will resist the corrosiveness of lead free solder alloys.