Desoldering Depaneling Tools, Fancort Industries
Supplier: Fancourt
NTR-2
76249-402EA 4320 USD
76249-402
Desoldering Depaneling Tools, Fancort Industries
Soldering Supplies Desoldering Tools
This affordable low volumn tab router features with A2 tool steel knives, foot switch operated and scrap collector.
  • Low automation
  • Low capacity and high strees level
Board is made of NR4 material.