PHS-1KBIR Preheater Set, JBC
Supplier: JBC TOOLS USA INC. TE
PHSE-1KB
76248-794EA 7717.92 USD
76248-794
PHS-1KBIR Preheater Set, JBC
Soldering Supplies Desoldering Tools
Complete system for preheating multilayer PCBs and ideal for repetitive soldering jobs.
  • Perfectly adapted to any type of PCBs
  • Three adjustable height positions
The infrared preheater PHS is used as a secondary heat source to reduce the amount of energy supplied by the main soldering tool, reducing thermal shock to components and PCBs.
Heating area is measured as 6.5×13.5 cm for one zone and 13×13.5 cm for two zones.
Delivery information: Consists of close loop thermocouple controlled infrared preheater unit and with a PCB support.