Tip Tinner, Lead Free, R & R Lotion
Supplier: R & R Lotion
TIP-2
76241-918CS 400.14 USD
76241-918
Tip Tinner, Lead Free, R & R Lotion
Soldering Supplies Desoldering Tools
All the major manufacturers of soldering tips have one thing in common, they all recommend that “Tin Soldering Tips”. I.C. Lead Free Tip Tinner has been formulated to prolong soldering tip life in the harsh environment of no clean and water soluble fluxes.