Glycerol Colorimetric Assay Kit, Cayman Chemical Company
Supplier: Cayman Chemical Company
10010755-96
75816-762EA 345.58 USD
75816-762
Glycerol Colorimetric Assay Kit, Cayman Chemical Company
Assays Protein Assays
Simple, reproducible, and sensitive tool for measurement of glycerol in plasma and serum.
  • Assay 40 samples in duplicate
  • Plate-based colorimetric measurement (530 – 550 nm)
Glycerol is the backbone of triglycerides and an important intermediate in energy metabolism being involved in both oxidative and synthetic processes. The circulating levels of glycerol and free fatty acids are considered to reflect lipolysis and are therefore useful parameters to evaluate in various conditions. The measurement of glycerol is also useful for correction of glycerol interference when measuring triglycerides in reference materials and in serum from patients with elevated glycerol concentrations. The assay employs a coupled enzymatic reaction system that yields a brilliant purple product with an absorbance maximum at 540 nm.