Protein Carbonyl Colorimetric Assay Kit, Cayman Chemical Company
Supplier: Cayman Chemical Company
10005020-96
75816-674EA 465.48 USD
75816-674
Protein Carbonyl Colorimetric Assay Kit, Cayman Chemical Company
Assays Protein Assays
Convenient colorimetric assay for measurement of oxidized proteins.
  • Measure protein carbonyl content in plasma, serum, urine, tissue homogenates, and cell lysates
  • Assay 24 samples in duplicate
  • Plate-based colorimetric measurement (360 – 385 nm)
The carbonyls of protein samples are derivatized using 2,4-dinitrophenylhydrazine. Formation of a Schiff base produces the corresponding hydrazone which can be analyzed spectrophotometrically at 360 – 385 nm. This assay can be used to measure oxidized protein in plasma, serum, cell lysates, and tissue homogenates.