Sulfuric acid ≥96%, Cleanroom® MB for the electronics industry

Supplier: KMG
Danger

64071
71009-532EA 4779.57 USD
71009-532
Sulfuric acid ≥96%, Cleanroom® MB for the electronics industry
Sulfuric acid

Sulfuric acid is used in semiconductor processing to remove residual organic contamination prior to metalization steps.

Store at less than 109⁰ F

Formula: H₂SO₄
MW: 98.08 g/mol
Boiling Pt: ∼335 °C (1013 hPa)
Melting Pt: 3 °C
Density: 1.84 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 1830
ADR: 8,II
Merck Index: 14,08974

Order Now

Specification Test Results

Assay (H2SO4) 95.00 to 97.00 %
Assay (H2O) 3.00 to 5.00 %
Color 10 APHA
Appearance Conforms
Substance Reducing KMnO4 Conforms
Chloride (Cl) 100 ppb
Nitrate (NO3) 100 ppb
Phosphate (PO4) 200 ppb
Aluminum (Al) 10.0 ppb
Antimony (Sb) 1.0 ppb
Arsenic (As) 1.0 ppb
Barium (Ba) 10.0 ppb
Beryllium (Be) 10.0 ppb
Bismuth (Bi) 10.0 ppb
Boron (B) 10.0 ppb
Cadmium (Cd) 10.0 ppb
Calcium (Ca) 10.0 ppb
Chromium (Cr) 10.0 ppb
Cobalt (Co) 5.0 ppb
Copper (Cu) 10.0 ppb
Gallium (Ga) 10.0 ppb
Germanium (Ge) 10.0 ppb
Gold (Au) 5.0 ppb
Iron (Fe) 10.0 ppb
Lead (Pb) 10.0 ppb
Lithium (Li) 10.0 ppb
Magnesium (Mg) 10.0 ppb
Manganese (Mn) 10.0 ppb
Molybdenum (Mo) 10.0 ppb
Nickel (Ni) 10.0 ppb
Niobium (Nb) 10.0 ppb
Potassium (K) 10.0 ppb
Silicon (Si) 10.0 ppb
Silver (Ag) 10.0 ppb
Sodium (Na) 10.0 ppb
Strontium (Sr) 10.0 ppb
Tantalum (Ta) 10.0 ppb
Thallium (Tl) 10.0 ppb
Tin (Sn) 10.0 ppb
Titanium (Ti) 10.0 ppb
Vanadium (V) 10.0 ppb
Zinc (Zn) 10.0 ppb
Zirconium (Zr) 10.0 ppb
0.5μ Particle Count 25 par/ml
1.0μ Particle Count 10 par/ml
10 par/ml

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR