Acetone ≥99.5%, Cleanroom® LP for the electronics industry

Supplier: KMG
Danger

Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone

200438
71009-508EA 4158.17 USD
71009-508
Acetone ≥99.5%, Cleanroom® LP for the electronics industry
Acetone
Formula: CH₃COCH₃
MW: 58.08 g/mol
Boiling Pt: 56.2 °C (1013 hPa)
Melting Pt: –95.4 °C
Density: 0.792 g/cm³ (20 °C)
Flash Pt: <–20 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00008765
CAS Number: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3,II
REACH: 01-2119471330-49
Merck Index: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.5 - 100%
Methanol 0 - 0.05%
Color APHA <10 AP
Acidity <0.3 µeq/g
Alkalinity <0.3 µeq/g
Water 0 - 0.5%
Aldehydes 0 - 0.002%
Residue After Evaporation <5000 ppb
Phosphate (PO₄) <50 ppb
Chloride (Cl) <200 ppb
Aluminum (Al) <50 ppb
Antimony (Sb) <5 ppb
Arsenic (As) <5 ppb
Barium (Ba) <50 ppb
Beryllium (Be) <50 ppb
Bismuth (Bi) <50 ppb
Boron (B) <50 ppb
Cadmium (Cd) <50 ppb
Calcium (Ca) <50 ppb
Chromium (Cr) <50 ppb
Cobalt (Co) <10 ppb
Copper (Cu) <10 ppb
Gallium (Ga) <50 ppb
Germanium (Ge) <50 ppb
Gold (Au) <50 ppb
Iron (Fe) <50 ppb
Lead (Pb) <50 ppb
Lithium (Li) <50 ppb
Magnesium (Mg) <50 ppb
Manganese (Mn) <50 ppb
Molybdenum (Mo) <50 ppb
Nickel (Ni) <10 ppb
Niobium (Nb) <50 ppb
Potassium (K) <50 ppb
Silicon (Si) <50 ppb
Silver (Ag) <50 ppb
Sodium (Na) <50 ppb
Strontium (Sr) <50 ppb
Tantalum (Ta) <50 ppb
Thallium (Tl) <50 ppb
Tin (Sn) <50 ppb
Titanium (Ti) <50 ppb
Vanadium (V) <50 ppb
Zinc (Zn) <50 ppb
Zirconium (Zr) <50 ppb
0.2 Micron Particle Count <200 Par/ml
0.3 Micron Particle Coun <200 Par/ml
0.5 Micron Particle Count <150 Par/ml
1.0 Micron Particle Count <25 Par/ml

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR