Xylene (mixture of isomers) ≥99.00%

Supplier: KMG

Synonyms: Dimethylbenzene

200554
71009-384CS 1014.58 USD
71009-384
Xylene (mixture of isomers) ≥99.00%
Xylene (mixture of isomers)

Xylene can be used as a negative resist developer in semiconductor applications. Xylene is also mixed with HMDS, then spun on to a wafer to aid in adhesive of photoresist to the silicon wafer.

Formula: C₆H₄(CH₃)₂
MW: 106.17 g/mol
Boiling Pt: 140 °C (1013 hPa)
Melting Pt: –34 °C
Density: 0.86…0.88 g/cm³ (20 °C)
Flash Pt: 25 °C
MDL Number: MFCD00077264
CAS Number: 1330-20-7
UN: 1307
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2deefc7b7b67c10e64003e243ff3216fad27cf7d&vid=9fa01ddafd4814d5f34266ae7933463a29d92d19&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (Xylene) 99.00
Assay (H2O) 0,20%
Color 10
Acidity 0.300µeq/g
Residue After Evaporation 5000 ppb
Chloride 3000 ppb
Phosphate 1000 ppb
Sulfur Compounds 30 ppb
Infared Conform
Appearance Conform
Sustance Darkening (by H2SO4) Conform
Aluminum 100 ppb
Antimony 10 ppb
Arsenic 10 ppb
Boron 100 ppb
Calcium 100 ppb
Chromium 100 ppb
Copper 10 ppb
Gold 100 ppb
Iron 100 ppb
Lead 100 ppb
Magnesium 100 ppb
Manganese 100 ppb
Nickel 10 ppb
Potassium 100 ppb
Sodium 100 ppb
Tin 100 ppb
Titanium 100 ppb
Zinc 100 ppb
1.0µ Particle Count 10 par/ml

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR