Potassium hydroxide 45% in aqueous solution, Cleanroom® LP LP for the electronics industry

Supplier: KMG
Danger

Synonyms: Potash caustic, Potassium hydrate, Caustic potash

64162
71009-346CS 803.43 USD
71009-346
Potassium hydroxide 45% in aqueous solution, Cleanroom® LP LP for the electronics industry
Potassium hydroxide

Potassium hydroxide (KOH) is used to etch silicon. The crystalline orientation of the silicon greatly affects the etch rate making potassium hydroxide an anisotropic etchant.

Storage: 32⁰ F - 120⁰ F

Formula: KOH
MW: 56.11 g/mol
Boiling Pt: 1320
Melting Pt: 360
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 1814
ADR: 8 II
Merck Index: 12,07806

Order Now

Specification Test Results

Assay (KOH) 45.00 47.00 %
Assay (H2O) 53.00 55.00 %
Color 10 APHA
Appearance Conforms
NH4OH Precipitate 0.005%
Nitrogen Compounds (as N ) 5,000 ppb
Insoluble Matter 50 ppm
Assay (K2CO3) 0.20%
Heavy Metals 5,000 ppb
Chloride (Cl) 50,000 ppb
Phosphate (PO4) 2,000 ppb
Sulfate (SO4) 10,000 ppb
Copper (Cu) 50,000 ppb
Iron (Fe) 2,000 ppb
Nickel (Ni) 5,000 ppb
Sodium (Na) 200,000 ppb


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR