Acetone, Cleanroom® MB

Supplier: KMG

Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone

200400
71009-318CS 420.17 USD
71009-318
Acetone, Cleanroom® MB
Acetone

can be used for drying laboratory glassware, removing greasy or oily contaminants from work benches, or as a general photoresist stripper or edgebead remover.

Formula: CH₃COCH₃
MW: 58.08 g/mol
MDL Number: MFCD00008765
CAS Number: 67-64-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR