Endura™ Competent Cells, Biosearch Technologies
Supplier: Lucigen
60240-1 60240-2 60241-1 60242-1 60242-2
71003-028EA 427.21 USD
71003-028 71003-030 71003-032 71003-036 71003-038
Endura™ Competent Cells, Biosearch Technologies
Competent Cells
With Lucigen’s Endura Competent cells, you can clone unstable sequences with exceptionally reliable results
Lucigen’s Endura Competent Cells are optimal for conserving clones that contain unstable sequences and maintaining unstable DNA. Endura Competent Cells are a commonly used strain for cloning sequences that suffer unwanted recombination events in other strains. Clones with inverted repeats or other sequences prone to recombination are commonly found in retroviral genes, and require cells such as Endura to be propagated stably.
Lucigen offers your choice of two packaging formats and noteably high efficiencies: over 1 × 10^8 cfu/µg (chem) or 1 × 10^10 cfu/µg (electro).
Lucigen’s Endura Competent Cells provide superior value and efficiency. Switch to Endura cells today and experience a higher level of efficiency and a savings in your cloning budget.
Available as chemically competent or electrocompetent cells, Endura Competent Cells are an excellent value and are direct replacements for Invitrogen Stbl3™ and Agilent SURE strains.
Ordering information: Endura Competent Cells include Control DNA and Recovery Medium, and are packaged as SOLOs (1 transformation per tube) or DUOs (2 transformations per tube) as indicated. Endura Electrocompetent cells are provided in DUOs format only. Recovery Medium is also available separately. The specified transformation efficiencies are with pUC DNA, unless indicated otherwise.