Chromium etchant CR-16

Supplier: KMG

210020
10803-036CS 510.39 USD
10803-036
Chromium etchant CR-16
Chromium etchant

CR-16, is a nitric acid based chrome etchant used for photolithography masks, semiconductor and MEMS processes. It is designed to provide minimum undercutting, maximum critical dimension control and repeatability.


Ordering information: Minimum ordering quantities may apply.

Order Now

Specification Test Results

Ceric Ammonium Nitrate 0.054 - 0.074%
Total Acidity 9.80 - 11.40%
pH 1
Specific Gravity 1.068 - 1.080

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR