Human Recombinant IL-23R (from CHO Cells)
Supplier: ADIPOGEN CORP MS
CHI-HF-21023R-C100
103692-504EA 484.62 USD
103692-504
Human Recombinant IL-23R (from CHO Cells)
Proteins and Peptides
Interleukin-23 receptor (IL-23R), is a member of the type I cytokine receptor family, type 2 subfamily. It is expressed by monocytes, Th1, Th0, NK and dendritic cells. Isoform 1 is specifically expressed in NK cells. IL-23 functions in innate and adaptive immunity and may participate in acute response to infection in peripheral tissues. IL-23 may be responsible for autoimmune inflammatory diseases and be important for tumorigenesis. IL-23R associates with IL-12RB1 to form the interleukin-23 receptor. It binds IL-23 and mediates T cells, NK cells and possibly certain macrophage/myeloid cells stimulation probably through activation of the Jak-Stat signaling cascade. Genetic variations in IL-23R are associated with inflammatory bowel disease type 17 (IBD17). IBD17 is a chronic, relapsing inflammation of the gastrointestinal tract with a complex etiology. Genetic variations in IL-23R also can cause susceptibility to psoriasis type 7.