Plerixafor (AMD3100)

Supplier: SELLECK CHEMICALS LLC MS

Synonyms: AMD 3100, Mezobil, JM 3100, Plerixafor, 1,4-Bis((1,4,8,11-tetraazacyclotetradecan-1-yl)methyl)benzene, 1,1'-(1,4-phenylenebis(methylene))bis(1,4,8,11-tetraazacyclotetradecane)octahydrochloride dihydrate, 1,4-Bis[(1,4,8,11-tetraazacyclotetradecan-1-yl)methyl]benzene

S8030-5MG S8030-10MG S8030-50MG
103548-030EA 181.54 USD
103548-030 103548-032 103548-034
Plerixafor (AMD3100)
Plerixafor (AMD3100)

Plerixafor (AMD3100) is a chemokine receptor antagonist for CXCR4 and CXCL12-mediated chemotaxis with IC50 of 44 nM and 5.7 nM in cell-free assays, respectively.

Formula: C₂₈H₅₄N₈
Storage Temperature: Freezer
MDL Number: MFCD05662218
CAS Number: 110078-46-1
UN: 3077
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR