HiLyte™ Fluor 532 acid fluorescent dye
Supplier: Anaspec Inc
HiLyte™ Fluor 532 dyes are new members of our proprietary HiLyte™ Fluor series with fluorescence excitation and emission maxima of ~545 nm and ~565 nm, respectively.
- Spectral Properties Abs/Em = 545/565 nm
- Solvent System Water
- Storage -20C desiccated and protected from light
The dyes are based on a new class of cyanine dyes developed by AnaSpec scientists. Unlike traditional cyanine dyes, HiLyte™ Fluor 532 dyes are not only highly water-soluble, pH-insensitive but also photostable. HiLyte™ Fluor 532 is suited for the frequency-doubled Nd:YAG laser line (Flow cytometry laser line: 532 nm; Microscopy laser line: 532 nm) and is a direct replacement for: Alexa Fluor® 532, Atto 532.
HiLyte™ Fluor 532 has Extinction Coefficient (M-1cm-1) = 171,000 and Fluorescence quantum yield = 0.26.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...