N,N-Dimethyl-6-propionyl-2-naphthylamine (Prodan)

Supplier: Anaspec Inc

Synonyms: 6-Propionyl-2-dimethylaminonaphthalene, 1-(6-(Dimethylamino)naphthalen-2-yl)propan-1-one, Prodan, 2-(Dimethylamino)-6-propionylnaphthalene

AS-88212
103011-376EA 220.69 USD
103011-376
N,N-Dimethyl-6-propionyl-2-naphthylamine (Prodan)
N,N-Dimethyl-6-propionyl-2-naphthylamine (Prodan)

Environment-sensitive dye for studying membranes and structures of proteins.


  • Spectral properties: Abs/Em = 363/497 nm
  • Solvent system: DMF
  • Storage: –20 °C desiccated and protected from light
  • Shipping condition: RT

Formula: C₁₅H₁₇NO
MW: 227.302 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00056615
CAS Number: 70504-01-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR