Tetramethylrhodamine ethyl ester perchlorate (TMRE) mitochondria dye, suitable for fluorescence

Supplier: Anaspec Inc
Warning

AS-88061
103011-368EA 285 USD
103011-368
Tetramethylrhodamine ethyl ester perchlorate (TMRE) mitochondria dye, suitable for fluorescence
Tetramethylrhodamine ethyl ester perchlorate (TMRE)

Used for measuring membrane potential of mitochondria; Fluorescence is less dependent on dye location.


  • Spectral Properties - Abs/Em = 549/574 nm

Formula: C₂₆H₂₇ClN₂O₇
Storage Temperature: Freezer
MDL Number: MFCD00467973
CAS Number: 115532-52-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR