JC-1 iodide salt

Supplier: Anaspec Inc

Synonyms: 5,5′,6,6′-Tetrachloro-1,1′,3,3′-tetraethylbenzimidazolocarbocyanine iodide, JC-1, 5,5′,6,6′-Tetrachloro-1,1′,3,3′-tetraethyl-imidacarbocyanine iodide, (E)-5,6-Dichloro-2-(3-(5,6-dichloro-1,3-diethyl-1,3-dihydro-2H-benzo[d]imidazol-2-ylidene)prop-1-en-1-yl)-1,3-diethyl-1H-benzo[d]imidazol-3-ium iodide, CBIC2(3), 1,1',3,3'-tetraethyl-5,5',6,6'-tetrachloroimidacarbocyanine iodide

AS-88060
103011-366EA 568.54 USD
103011-366
JC-1 iodide salt
JC-1 iodide salt

Widely used for measuring membrane potential of mitochondria; 585/520 Fluorescence ratio increases upon cell hyper-polarization.


  • Storage: -20°C desiccated and protected from light
  • Spectral Properties: Abs/Em = 514/529 nm
  • Solvent System: DMSO

Formula: C₂₅H₂₇Cl₄IN₄
Storage Temperature: Freezer
MDL Number: MFCD00188475
CAS Number: 47729-63-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR