Dihydrorhodamine 123

Supplier: Anaspec Inc

Synonyms: Methyl 2-(3,6-diamino-9H-xanthen-9-yl)benzoate, 2-(3,6-Diamino-9H-xanthene-9-yl)-benzoic acid methyl ester

AS-85711
103011-336EA 204.62 USD
103011-336
Dihydrorhodamine 123
Dihydrorhodamine 123

Generic substrate for fluorimetric detection of oxidases (including peroxidase) in mitochondria


  • Spectral properties: Abs/Em = 507/529 nm
  • Solvent system: DMSO
  • Storage Temperature: -20°C desiccated and protected from light
  • Shipping condition: RT

Formula: C₂₁H₁₈N₂O₃
MW: 346.386 g/mol
Storage Temperature: Freezer
MDL Number: MFCD04221428
CAS Number: 109244-58-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR