Gelatin, FITC (Fluorescein Isothiocyanate)
Supplier: Anaspec Inc
AS-85145
103011-298EA 201.69 USD
103011-298
Gelatin, FITC (Fluorescein Isothiocyanate)
Proteins and Peptides
This gelatin is heavily labeled with FITC, and the conjugate is a highly quenched gelatinase/collagenase substrate. It is efficiently digested by gelatinases and collagenases, releasing brightly fluorescent peptides. The increase in fluorescence upon digestion is proportional to proteolytic activity. Longer incubation may increase its sensitivity for detecting proteases.
  • Absorbance: Abs/Em = 492/515 nm