3,3'-Dipropylthiadicarbocyanine iodide [DiSC3(5)] ≥95% (by HPLC)

Supplier: Anaspec Inc

Synonyms: DiSC3(5), 3-Propyl-2-[5-[3-propyl-2(3H)-benzothiazolylidene]-1,3-pentadienyl]benzothiazolium iodide, 5-(3-Propylbenzothiazol-2-ylidene)-1-(3-propylbenzothiazolium-2-yl)-1,3-pentadiene odide, 3,3'-Dipropylthiadicarbocyanine iodide

AS-84923
103011-276EA 537.85 USD
103011-276
3,3'-Dipropylthiadicarbocyanine iodide [DiSC3(5)] ≥95% (by HPLC)
3,3'-Dipropylthiadicarbocyanine iodide [DiSC3(5)]

Accumulate in cells on hyperpolarized membranes

Formula: C₂₅H₂₇IN₂S₂
MW: 546.53 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00075033
CAS Number: 53213-94-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR