Di-4-ANEPPS membrane stain, for histology

Supplier: Anaspec Inc

Synonyms: 4-(2-(6-(Dibutylamino)-2-naphthalenyl)ethenyl)-1-(3-sulfopropyl)pyridinium hydroxide inner salt, 4-(2-(6-(Dibutylamino)-2-naphthalenyl)ethenyl)-1-(3-sulphopropyl)pyridinium hydroxide inner salt

AS-84723
103011-252EA 838.92 USD
103011-252
Di-4-ANEPPS membrane stain, for histology
Di-4-ANEPPS

Fluorescence response less susceptible to cell and tissue types


  • Abs/Em = 496/705 nm

Formula: C₂₈H₃₆N₂O₃S
MW: 480.66 g/mol
Storage Temperature: Freezer
MDL Number: MFCD03453609
CAS Number: 90134-00-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR