HiLyte™ Fluor 405 acid, SE fluorescent dye
Supplier: Anaspec Inc
AS-89317-1
103011-236EA 280.62 USD
103011-236
HiLyte™ Fluor 405 acid, SE fluorescent dye
HiLyte™ Fluor
HiLyte™ Fluor 405 is a blue fluorescent dye with Ex/Em = 404/428 nm, spectrally similar to Alexa Fluor™ 405 and DyLight Fluor™ 405 dyes.
  • Abs/Em = 404/428 nm
Conjugates of HiLyte™ Fluor 405 are well excited by the 407 nm spectral line of the Krypton laser and the 408 nm line of the violet laser diode. HiLyte™ Fluor 405 acid, SE is an amine reactive reagent for labeling peptides, proteins and antibodies.
Extinction Coefficient (M-1cm-1):35,000
Fluorescence quantum yield: 0.54