Calcein AM ≥95% (by HPLC), Ultra Pure Grade fluorescent dye

Supplier: Anaspec Inc

Synonyms: Calcein-AM, Calcein O,O′-diacetate tetrakis(acetoxymethyl) ester, Calcein acetoxymethyl ester, Tetrakis(acetoxymethyl) 2,2',2'',2'''-(((3',6'-diacetoxy-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-2',7'-diyl)bis(methylene))bis(azanetriyl))tetraacetate

AS-89204 AS-89202
103011-230EA 529.08 USD
103011-230 103011-398
Calcein AM ≥95% (by HPLC), Ultra Pure Grade fluorescent dye
Calcein AM

Calcein, AM is a cell-permeant and non-fluorescent compound that is widely used for determining cell viability


  • Spectral Properties: Abs/Em = 494/517 nm
  • Storage: -20°C desiccated and protected from light


In live cells the non-fluorescent calcein, AM is hydrolyzed by intracellular esterases into the strongly green fluorescent anion calcein. The fluorescent calcein is well-retained in the cytoplasm in live cells. Hela cells stained with AnaSpec’s Calcein AM fluorescent dye (Cat# AS-89201, AS-89202, AS-89203 and AS-89204).

Formula: C₄₆H₄₆N₂O₂₃
MW: 994.86 g/mol
Storage Temperature: Freezer
MDL Number: MFCD05861516
CAS Number: 148504-34-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR