Fura 2-AM ≥95% (by HPLC)

Supplier: Anaspec Inc

Synonyms: FURA-2 pentakis(acetoxymethyl) ester, 1-[2-(5-Carboxyoxazol-2-yl)-6-aminobenzofuran-5-oxy]-2-(2'-amino-5'-methyl-phenoxy)ethane-N,N,N',N'-tetraacetic acid, pentaacetoxymethyl ester, FURA-2 acetoxymethyl ester, Fura-2 AM Ester

AS-84015
103011-172EA 632.85 USD
103011-172
Fura 2-AM ≥95% (by HPLC)
Fura 2-AM

Excitation-ratiometric fluorescent indicator for quantifying intracellular Ca2+ concentration


  • Storage: -20°C desiccated and protected from light
  • Spectral Properties: Abs/Em = 363/512 nm
  • Solvent System: DMSO

Formula: C₄₄H₄₇N₃O₂₄
MW: 1001.857 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00036976
CAS Number: 108964-32-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR