Phthalaldehyde ≥95% (by HPLC), Ultra Pure Grade

Supplier: Anaspec Inc

Synonyms: 1,2-Phthalic dicarboxaldehyde, Phthaldialdehyde, o-Phthalaldehyde, OPA, Phthaldicarboxaldehyde, Benzene-1,2-dicarboxaldehyde

AS-83012
103011-116EA 190 USD
103011-116
Phthalaldehyde ≥95% (by HPLC), Ultra Pure Grade
Phthalaldehyde

Fluorogenic indicator for primary amino group; Widely used for quantitation of peptides and proteins.

Formula: C₈H₆O₂
MW: 134.13 g/mol
Boiling Pt: 266
Flash Pt: 132
Storage Temperature: Freezer
MDL Number: MFCD00003335
CAS Number: 643-79-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR