2,3-Naphthalenedialdehyde (NDA) ≥95% (by HPLC), Ultra Pure Grade

Supplier: Anaspec Inc

AS-83010
103011-112EA 233.85 USD
103011-112
2,3-Naphthalenedialdehyde (NDA) ≥95% (by HPLC), Ultra Pure Grade
2,3-Naphthalenedialdehyde (NDA)

Fluorogenic indicator for primary amino group; Widely used for quantitation of peptides and proteins.

Formula: C₁₂H₈O₂
Storage Temperature: Freezer
MDL Number: MFCD00016618
CAS Number: 7149-49-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR