D-(-)-Luciferin potassium salt ≥95% (by HPLC), Ultrapure

Supplier: Anaspec Inc

AS-82252
103011-096EA 210.46 USD
103011-096
D-(-)-Luciferin potassium salt ≥95% (by HPLC), Ultrapure
D(-)-Luciferin potassium salt

Luminescent substrate for firefly luciferase.


  • Spectral properties: Abs/Em = 328/532 nm
  • Solvent system: Water
  • Storage Temperature: -20°C desiccated and protected from light
  • Shipping condition: RT

Formula: C₁₁H₇KN₂O₃S₂
Storage Temperature: Freezer
MDL Number: MFCD00044928
CAS Number: 115144-35-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR