D-(-)-Luciferin free acid, Ultrapure

Supplier: Anaspec Inc

Synonyms: D-Luciferin, (S)-2-(6-Hydroxybenzo[d]thiazol-2-yl)-4,5-dihydrothiazole-4-carboxylic acid

AS-82250
103011-092EA 201.69 USD
103011-092
D-(-)-Luciferin free acid, Ultrapure
D(-)-Luciferin

Luminescent substrate for firefly luciferase.


  • Spectral properties: Abs/Em = 328/532 nm
  • Solvent system: water
  • Physical State: Solid
  • Storage Temperature: -20°C desiccated and protected from light
  • Shipping condition: RT

Formula: C₁₁H₈N₂O₃S₂
MW: 280.3228 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00042929
CAS Number: 2591-17-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR