5-FITC cadaverine

Supplier: Anaspec Inc

Synonyms: 5-((5-Aminopentyl)thioureidyl)fluorescein

AS-81504
103011-024EA 232.38 USD
103011-024
5-FITC cadaverine
5-FITC cadaverine

Fluorescein cadaverine is a popular fluorescent transglutaminase substrate to label proteins by transaminidation. It has also been used to prepare other small fluorescent biomolecules via amidation or reductive amination.


  • Spectral properties: Abs/Em = 492/516 nm
  • Solvent system: DMF or DMSO
  • Storage Temperature: -20°C desiccated and protected from light
  • Shipping condition: Blue ice

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR