Tetramethylrhodamine-5-(-6) C2 maleimide
Supplier: Anaspec Inc
AS-81441-25
AS-81441-5
103011-000EA
1074.23
USD
103011-000
103011-002
Tetramethylrhodamine-5-(-6) C2 maleimide
Tetramethylrhodamine-5-(-6) C2 maleimide
Tetramethylrhodamine-5-(and-6) C2 maleimide is a good alternative to tetramethylrhodamine-5-(and-6)-maleimide with excitation/emission wavelength at 544/572 nm. Its spectral characteristics are very close to those of tetramethylrhodamine-5-(and-6)-maleimide (+2 nm). It is 40% less expensive compared to tetramethylrhodamine-5-(and-6)-maleimide, and can be used for thiol modification of proteins, antibodies and peptides.
- Spectral properties: Abs/Em = 544/572nm
- Solvent system: DMF or DMSO
- Storage Temperature: -20°C desiccated and protected from light
- Shipping condition: Blue ice
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...