Bromobimane (mBBr), Ultra Pure Grade

Supplier: Anaspec Inc

AS-81433
103010-996EA 203.15 USD
103010-996
Bromobimane (mBBr), Ultra Pure Grade
Bromobimane (mBBr)

mBBr is one of the most popular thiol-reactive fluorescent tags, and is widely used to detect various thiol-containing biomolecules such as glutathione in cells. It is fluorogenic upon reacting with thiol-containing molecules.


  • Storage: -20°C desiccated and protected from light
  • Spectral Properties: Abs/Em = 395/490 nm
  • Solvent System: DMSO

Formula: C₁₀H₁₁BrN₂O₂
Storage Temperature: Freezer
MDL Number: MFCD00036951
CAS Number: 71418-44-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR