HiLyte™ Fluor 750 hydrazide fluorescent dye

Supplier: Anaspec Inc

AS-81268
103010-950EA 470.62 USD
103010-950
HiLyte™ Fluor 750 hydrazide fluorescent dye
HiLyte™ Fluor

HiLyte™ Fluor 750 hydrazide is a carbonyl-reactive fluorescent labeling dye. It can be used for labeling glycoproteins such as HRP.


  • Spectral Properties Abs/Em = 753/782 nm
  • Solvent System water or DMF
  • Storage -20°C desiccated and protected from light


Spectrally similar to Cy7 dye, HiLyte™ Fluor 750 hydrazide is the longest wavelength carbonyl-reactive HiLyte™ Fluor dye currently available. Its fluorescence emission maximum at 782 nm is well separated from commonly used far-red fluorophores such as HiLyte™ Fluor 647, HiLyte™ Fluor 680 or allophycocyanin (APC), facilitating multicolor analysis.


Extinction Coefficient (M-1cm-1): 275,000
Fluorescence quantum yield: 0.12
Fluorescence Life Time (ns): 0.7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR