HiLyte™ Fluor 750 amine fluorescent dye
Supplier: Anaspec Inc
HiLyte™ Fluor 750 amine is a carbonyl-reactive fluorescent labeling dye. Spectrally similar to Cy7 dye, HiLyte™ Fluor 750 amine is the longest-wavelength carbonyl-reactive HiLyte™ Fluor dye currently available.
- Spectral Properties Abs/Em = 754/778 nm
- Solvent System water
- Storage -20°C desiccated and protected from light
Its fluorescence emission maximum at 778 nm is well separated from commonly used far-red fluorophores such as HiLyte™ Fluor 647, HiLyte™ Fluor 680 or allophycocyanin (APC), facilitating multicolor analysis. With a peak excitation at ~754 nm, conjugates of HiLyte™ Fluor 750 dyes are well excited by a xenon-arc lamp or dye-pumped lasers operating in the 720–750 nm range.
Extinction Coefficient (M-1cm-1): 275,000
Fluorescence quantum yield: 0.12
Fluorescence Life Time (ns): 0.7
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...