HiLyte™ Fluor 555 acid fluorescent dye
Supplier: Anaspec Inc
AS-81250
103010-932EA
1376.77
USD
103010-932
HiLyte™ Fluor 555 acid fluorescent dye
HiLyte™ Fluor
Spectra of HiLyte™ Fluor 555 conjugates are slightly red-shifted compared to those of Cy3 dyes, resulting in an optimal match to filters designed for Cy3 dyes.
- Spectral Properties Abs/Em = 550/566 nm
- Solvent System water
- Storage -20°C desiccated and protected from light
HiLyte™ Fluor 555 is more photostable than Cy3, providing researchers with additional time for image capture. Additionally, our in-house data indicated that HiLyte™ Fluor 555 is superior to Cy3 in fluorescence polarization-based assays.
Extinction Coefficient (M-1cm-1): 150,000
Fluorescence quantum yield: 0.10
Fluorescence Life Time (ns): 0.3
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...