HiLyte™ Fluor 488 hydrazide fluorescent dye
Supplier: Anaspec Inc
AS-81163
103010-880EA
333.23
USD
103010-880
HiLyte™ Fluor 488 hydrazide fluorescent dye
HiLyte™ Fluor
HiLyte™ Fluor 488 hydrazide is a carbonyl-reactive fluorescent labeling dye.
- Spectral Properties Abs/Em = 503/528 nm
- Solvent System DMF or DMSO
- Storage -20°C desiccated and protected from light
It can be used for labeling glycoproteins such as HRP. Fluorescence of HiLyte™ Fluor 488 fluorophore is independent of pH from 4 to 10. This pH insensitivity is a major improvement over fluorescein, which emits its maximum fluorescence only above pH 10.
Extinction Coefficient (M-1cm-1): 70,000
Fluorescence quantum yield: 0.91
Fluorescence Life Time (ns): 4.1
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...