5(6)-Carboxytetramethylrhodamine succinimidyl ester [5(6)-TAMRA SE]

Supplier: Anaspec Inc

Synonyms: NHS-Rhodamine, NHS-Rhoda, TAMRA SE, 5 and 6 isomers, TMR-SE, 5(6)-Carboxytetramethylrhodamine N-succinimidyl ester, 5(6)-TAMRA N-succinimidyl ester

AS-81124 AS-81124-01
103010-844EA 249.92 USD
103010-844 103010-846
5(6)-Carboxytetramethylrhodamine succinimidyl ester [5(6)-TAMRA SE]
5(6)-Carboxytetramethylrhodamine succinimidyl ester [5(6)-TAMRA SE]

Tetramethylrhodamine (TMR) is one of the fluorophores most often used for preparing peptide, protein, nucleotide and nucleic acid conjugates, especially fluorescent antibodies and avidin derivatives used in immunochemistry. TMR dyes have been widely used as acceptors for FAM fluorophores in a variety of FRET studies. The succinimidyl esters of 5-TAMRA, 6-TAMRA or the mixed isomers are the primary labeling reagents.


  • Spectral properties: Abs/Em = 546/575 nm
  • Solvent system: DMF or DMSO
  • Appearance: Dark red solid
  • Storage Temperature: -20°C desiccated and protected from light
  • Shipping condition: Blue ice

Formula: C₂₉H₂₅N₃O₇
MW: 527.52 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00077324
CAS Number: 150408-83-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR