6-TAMRA special formulation

Supplier: Anaspec Inc

Synonyms: 6-Carboxytetramethylrhodamine, 2-(3,6-Bis(dimethylamino)xanthylium-9-yl)-4-carboxybenzoate

AS-81122 AS-81122-01
103010-840EA 182.69 USD
103010-840 103010-842
6-TAMRA special formulation
6-TAMRA

6-TAMRA is the other purified single isomer of 5(6)-TAMRA. It is predominantly used for nucleotide labeling.


  • Spectral properties: Abs/Em = 541/568 nm
  • Solvent system: DMF or DMSO
  • Storage Temperature: -20°C desiccated and protected from light
  • Shipping condition: Blue ice

Formula: C₂₅H₂₂N₂O₅
Storage Temperature: Freezer
MDL Number: MFCD01073638
CAS Number: 91809-67-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR