5-Carboxytetramethylrhodamine (5-TAMRA) special formulation
Supplier: Anaspec Inc
Synonyms:
5-TAMRA, 5-Carboxytetramethylrhodamine, 2-(3,6-Bis(dimethylamino)xanthylium-9-yl)-5-carboxybenzoate, 2-[3,6-Bis(dimethylamino)xanthen-10-ium-9-yl]-5-carboxybenzoate, TAMRA, 5-isomer
AS-81121
AS-81121-01
AS-81121-1
103010-834EA
182.69
USD
103010-834
103010-836
103010-838
5-Carboxytetramethylrhodamine (5-TAMRA) special formulation
5-Carboxytetramethylrhodamine (5-TAMRA)
5-TAMRA is the purified single isomer of 5(6)-TAMRA. It is preferred for some complicated biological applications where reproducibility is more critical than material cost since the minor positional difference between 5-TAMRA and 6-TAMRA might affect some biological properties of the underlying conjugates.
- Spectral properties: Abs/Em = 541/568 nm
- Solvent system: DMF or DMSO
- Storage Temperature: -20°C desiccated and protected from light
- Shipping condition: Blue ice
Formula:
C₂₅H₂₂N₂O₅ Storage Temperature: Freezer |
MDL Number:
MFCD00269769 CAS Number: 91809-66-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...