5(6)-TAMRA special formulation

Supplier: Anaspec Inc

Synonyms: TAMRA 5 and 6-isomer, Tetramethylrhodamine-5-(and-6)-maleimide, 5(6)-Carboxy-tetramethylrhodamine, 9-[2,4(or 2,5)-Dicarboxyphenyl]-3,6-bis(dimethylamino)-xanthylium inner salt

AS-81120 AS-81120-01 AS-81120-1
103010-826EA 54.08 USD
103010-826 103010-828 103010-830
5(6)-TAMRA special formulation
5(6)-TAMRA

5(6)-TAMRA is the mixture of two carboxy tetramethylrhodamine (TMR) isomers. It is used to modify amino and hydroxy groups using EDC-mediated couplings when there are difficulties in using 5(6)-TAMRA, SE. TAMRA is one of the most popular fluorophores used in various bioconjugations.


  • Spectral properties: Abs/Em = 541/565 nm
  • Solvent system: DMF or DMSO
  • Physical State: Solid
  • Storage Temperature: -20°C desiccated and protected from light
  • Shipping condition: Blue ice

Formula: C₂₅H₂₂N₂O₅
MW: 430.45 g/mol
Storage Temperature: Freezer
MDL Number: MFCD16661194
CAS Number: 98181-63-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR