6-Carboxy-X-rhodamine succinimidyl ester (6-ROX SE)

Supplier: Anaspec Inc

Synonyms: 6-ROX, SE, 6-Carboxy-X-rhodamine, NHS ester, 6-ROX N-succinimidyl ester, Carboxy-X-rhodamine succinimidyl ester, 6-isomer

AS-81115-
103010-824EA 356.62 USD
103010-824
6-Carboxy-X-rhodamine succinimidyl ester (6-ROX SE)
6-Carboxy-X-rhodamine succinimidyl ester (6-ROX SE)

6-ROX, SE is the other purified single isomer of 5(6)-ROX, SE. It appears that 5-ROX is more often used than 6-ROX for labeling peptides and proteins. 6-ROX is predominately used for labeling nucleotides and sequencing nucleic acids.


  • Spectral properties: Abs/Em = 575/602 nm
  • Solvent system: DMF or DMSO
  • Storage Temperature: -20°C desiccated and protected from light
  • Shipping condition:

Formula: C₃₇H₃₃N₃O₇
MW: 631.685 g/mol
Storage Temperature: Freezer
CAS Number: 216699-36-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR