5(6)-Carboxy-X-rhodamine

Supplier: Anaspec Inc

Synonyms: 5(6)-ROX

AS-81110
103010-816EA 235.31 USD
103010-816
5(6)-Carboxy-X-rhodamine
5(6)-Carboxy-X-rhodamine

ROX dyes have longer excitation and emission wavelengths than the other ‘conventional’ rhodamines. These dyes are used to label peptides, proteins, and other biological ligands. 5-ROX, 6-ROX and their mixture 5(6)-ROX are used to label biomolecules by EDC-mediated reactions.


  • Spectral properties: Abs/Em = 568/591 nm
  • Solvent system: DMF or DMSO
  • Storage Temperature: -20°C desiccated and protected from light
  • Shipping condition: Blue ice

Formula: C₃₃H₃₀N₂O₅
MW: 534.612 g/mol
Storage Temperature: Freezer
MDL Number: MFCD01862979
CAS Number: 198978-94-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR