6-HEX, acid

Supplier: Anaspec Inc

Synonyms: 6-carboxy-2',4,4',5',7,7'-hexachlorofluorescein

AS-81019
103010-792EA 355.15 USD
103010-792
6-HEX, acid
6-HEX, acid

6-HEX is a popular amino-reactive fluorescent probe that is widely used in nucleic acid sequencing and related research. It can also be used to label peptides and oligonucleotides.


  • Spectral properties: Abs/Em = 533/550 nm
  • Solvent system: DMF or DMSO
  • Storage Temperature: -20°C desiccated and protected from light
  • Shipping condition: RT

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR