5(6)-Carboxyfluorescein N-hydroxysuccinimide ester (5/6-FAM SE)
Supplier: Anaspec Inc
Synonyms:
NHS-Fluorescein, FLUOS, 5(6)-FAM SE
Although FITC reagents have been more often used to prepare fluoresceinated bioconjugates, the low stability of FITC bioconjugates makes some researchers use amine-reactive succinimidyl esters of carboxyfluorescein (commonly called FAM) in bioconjugations. FAM reagents give carboxamides that are more resistant to hydrolysis. We have shown that FAM reagents require less stringent reaction conditions and give better conjugation yields, and the resulted conjugates have superior stability. We noted that FITC labeled peptides tend to deteriorate more quickly than the corresponding FAM conjugates.
- Spectral properties: Abs/Em = 494/519 nm
- Solvent system: DMF or DMSO
- Appearance: Orange Powder
- Solubility: 1 mmol in 1ml DMF
- Storage Temperature: -20°C desiccated and protected from light
- Shipping condition: Blue ice
Formula:
C₂₅H₁₅NO₉ Storage Temperature: Freezer |
MDL Number:
MFCD04119713 CAS Number: 117548-22-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...