Staphylococcus aureus Recombinant Protein A (from E. coli), HiLyte Fluor® 647
Supplier: Anaspec Inc
AS-72237
103010-696EA 282.08 USD
103010-696
Staphylococcus aureus Recombinant Protein A (from E. coli), HiLyte Fluor® 647
Proteins and Peptides
Protein A-HiLyte™ Fluor 647 Conjugate can be used as a universal reagent to detect primary antibodies in IHC from many species including rabbit, human, some mouse IgG isotypes, and others. Fluorescence (near-infrared) Excitation/Emission wavelength: 649 nm/674 nm.
AnaSpec's recombinant Protein A consists of only IgG binding domains and was expressed using a recombinant bacterial expression system. Its apparent molecular weight is approximately 30,000 Da compared to 42,000 Da for the native Protein A that is found in Staphylococcus aureus.
Protein A conjugates are widely used as labeled reagents for a variety of experiments including immunoprecipitation, antibody detection and purification and assay development